accessibility menu, dialog, popup

UserName

CAS RN 16024-82-1

IUPAC Name: methyl 2-isothiocyanatobenzoate
Synonyms: methyl 2-isothiocyanatobenzoate
Molecular Weight (g/mol): 193.22
Molecular Formula: C9H7NO2S
InChi Key: UNXVHBOJSCWVCD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results