missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluoro-5-nitrobenzoate, 98%

Catalog No. p-7053912
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6167606 5 g
AAH6167614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6167606 Supplier Thermo Scientific Chemicals Supplier No. H6167606

CAS: 2965-22-2 | C8H6FNO4 | 199.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2965-22-2
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.137
MDL Number MFCD06203689
InChI Key JZLONOOYIXEAHM-UHFFFAOYSA-N
Synonym methyl 2-fluoro-5-nitrobenzenecarboxylate, 2-fluoro-5-nitrobenzoic acid methyl ester, 5-nitro-2-fluorobenzoic acid methyl ester, benzoicacid, 2-fluoro-5-nitro-, methyl ester, benzoic acid, 2-fluoro-5-nitro-, methyl ester, methyl 2-fluoro-5-nitro-benzoate, acmc-209ha6, ksc563m1n, methyl2-fluoro-5-nitrobenzoate, 4-fluoro-3-methoxycarbonyl nitrobenzene
PubChem CID 11217860
IUPAC Name methyl 2-fluoro-5-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])F

Specifications

Quantity 5 g
Formula Weight 199.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-fluoro-5-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only