CAS RN 2965-22-2
IUPAC Name:
methyl 2-fluoro-5-nitrobenzoate
Synonyms:
methyl 2-fluoro-5-nitrobenzoate
Molecular Weight (g/mol):
199.14
Molecular Formula:
C8H6FNO4
InChi Key:
JZLONOOYIXEAHM-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(F)C=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Methyl 2-fluoro-5-nitrobenzoate, 98%
Methyl 2-fluoro-5-nitrobenzoate, 98%
CAS: 2965-22-2 Molecular Formula: C8H6FNO4 Molecular Weight (g/mol): 199.137 MDL Number: MFCD06203689 InChI Key: JZLONOOYIXEAHM-UHFFFAOYSA-N Synonym: methyl 2-fluoro-5-nitrobenzenecarboxylate,2-fluoro-5-nitrobenzoic acid methyl ester,5-nitro-2-fluorobenzoic acid methyl ester,benzoicacid, 2-fluoro-5-nitro-, methyl ester,benzoic acid, 2-fluoro-5-nitro-, methyl ester,methyl 2-fluoro-5-nitro-benzoate,acmc-209ha6,ksc563m1n,methyl2-fluoro-5-nitrobenzoate,4-fluoro-3-methoxycarbonyl nitrobenzene PubChem CID: 11217860 IUPAC Name: methyl 2-fluoro-5-nitrobenzoate SMILES: COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])F