missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluoro-4-hydroxybenzoate, 98%

Catalog No. AAH6167503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6167503 1 g
AAH6167506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6167503 Supplier Thermo Scientific Chemicals Supplier No. H6167503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 197507-22-5 | C8H7FO3 | 170.139 g/mol

Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 197507-22-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD09038282
InChI Key WYSPMXSNCAFCFV-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid methyl ester, 2-fluoro-4-hydroxybenzoic acid methyl ester, benzoic acid, 2-fluoro-4-hydroxy-, methyl ester, methyl-2-fluoro4-hydroxybenzoate, methyl2-fluoro-4-hydroxybenzoate, methyl-2-fluoro-4-hydroxybenzoate, methyl 2-fluoro-4-hydroxy-benzoate, 2-fluoro-4-hyroxybenzoic acid methyl ester
PubChem CID 22463192
IUPAC Name methyl 2-fluoro-4-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)F

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material Methyl 2-fluoro-4-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only