accessibility menu, dialog, popup

UserName

CAS RN 197507-22-5

IUPAC Name: methyl 2-fluoro-4-hydroxybenzoate
Synonyms: methyl 2-fluoro-4-hydroxybenzoate
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: WYSPMXSNCAFCFV-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results