missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromohexanoate 98.0+%, TCI America™

$33.56 - $139.81

Chemical Identifiers

CAS 5445-19-2
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00045038
InChI Key YGLPDRIMFIXNBI-UHFFFAOYSA-N
Synonym 2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
PubChem CID 95578
IUPAC Name methyl 2-bromohexanoate
SMILES CCCCC(C(=O)OC)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M237825G
View Documents
TCI America
M237825G
25 g
Each for $33.56
Only null left
 
M2378250G
View Documents
TCI America
M2378250G
250 g
Each for $139.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5445-19-2
209.083
YGLPDRIMFIXNBI-UHFFFAOYSA-N
95578
CCCCC(C(=O)OC)Br
C7H13BrO2
MFCD00045038
2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
methyl 2-bromohexanoate

Specifications

5445-19-2
94°C
MFCD00045038
2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
CCCCC(C(=O)OC)Br
209.083
209.08
Liquid
Colorless
C7H13BrO2
25 g
YGLPDRIMFIXNBI-UHFFFAOYSA-N
methyl 2-bromohexanoate
95578
≥98.0% (GC)
Methyl 2-Bromohexanoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3662

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'