accessibility menu, dialog, popup

UserName

H3CBrOOCH3

CAS RN 5445-19-2

IUPAC Name: methyl 2-bromohexanoate
Synonyms: methyl 2-bromohexanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: YGLPDRIMFIXNBI-UHFFFAOYNA-N
SMILES: CCCCC(Br)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results