missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromo-5-methoxybenzoate 98.0+%, TCI America™

Catalog No. M19345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19345G 5 g
M193425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19345G Supplier TCI America Supplier No. M19345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35450-36-3
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00051594
InChI Key VRTQLDFCPNVQNT-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzoic acid methyl ester, 2-bromo-5-methoxy-benzoic acid methyl ester, methyl-2-bromo-5-methoxybenzoate, benzoic acid, 2-bromo-5-methoxy-, methyl ester, pubchem3965, acmc-1ahhz, methyl 6-bromo-m-anisate, rarechem al bf 1111, attercop-chm at108617, methyl 2-bromo-5-methoxylbenzoate
PubChem CID 2776849
IUPAC Name methyl 2-bromo-5-methoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC=C1Br

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromo-5-methoxybenzoate