accessibility menu, dialog, popup

UserName

CAS RN 35450-36-3

IUPAC Name: methyl 2-bromo-5-methoxybenzoate
Synonyms: methyl 2-bromo-5-methoxybenzoate
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: VRTQLDFCPNVQNT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=CC=C1Br
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results