missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-5-methoxybenzoate, 98%

Catalog No. p-7022503
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1532206 5 g
AAA1532214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1532206 Supplier Thermo Scientific Chemicals Supplier No. A1532206
Only null left

CAS: 35450-36-3 | C9H9BrO3 | 245.07 g/mol

Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Avoid contact with strong oxidizing agents. Store in a cool and dark place. Protect from direct light.

Chemical Identifiers

CAS 35450-36-3
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00051594
InChI Key VRTQLDFCPNVQNT-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzoic acid methyl ester, 2-bromo-5-methoxy-benzoic acid methyl ester, methyl-2-bromo-5-methoxybenzoate, benzoic acid, 2-bromo-5-methoxy-, methyl ester, pubchem3965, acmc-1ahhz, methyl 6-bromo-m-anisate, rarechem al bf 1111, attercop-chm at108617, methyl 2-bromo-5-methoxylbenzoate
PubChem CID 2776849
IUPAC Name methyl 2-bromo-5-methoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC=C1Br

Specifications

Density 1.52
Boiling Point 271°C
Flash Point >110°C (230°F)
Refractive Index 1.564
Quantity 5 g
Beilstein 2641378
Solubility Information Not miscible in water.
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 2-bromo-5-methoxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only