missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-4-methylbenzoate, 98%

Catalog No. p-7054743
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6314403 1 g
AAH6314406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6314403 Supplier Thermo Scientific Chemicals Supplier No. H6314403

missing translation for 'validMainframeMsg'

CAS: 87808-49-9 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87808-49-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD03840737
InChI Key DJTUYAFJAGLQCK-UHFFFAOYSA-N
Synonym methyl2-bromo-4-methylbenzoate, 2-bromo-4-methylbenzoic acid methyl ester, benzoic acid, 2-bromo-4-methyl-, methyl ester, pubchem14035, acmc-209qq7, ksc495m0h, methyl 2-bromo-4-methyl-benzoate, 2-bromo-4-methyl-benzoic acid methyl ester
PubChem CID 21779704
IUPAC Name methyl 2-bromo-4-methylbenzoate
SMILES CC1=CC(=C(C=C1)C(=O)OC)Br

Specifications

Quantity 1 g
Formula Weight 229.07
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-bromo-4-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only