accessibility menu, dialog, popup

UserName

CH3OOCH3Br

CAS RN 87808-49-9

IUPAC Name: methyl 2-bromo-4-methylbenzoate
Synonyms: methyl 2-bromo-4-methylbenzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: DJTUYAFJAGLQCK-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Br)C=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results