missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Benzoylbenzoate 98.0+%, TCI America™
$57.79 - $380.27
Chemical Identifiers
| CAS | 606-28-0 |
|---|---|
| Molecular Formula | C15H12O3 |
| Molecular Weight (g/mol) | 240.258 |
| MDL Number | MFCD00017187 |
| InChI Key | NQSMEZJWJJVYOI-UHFFFAOYSA-N |
| Synonym | Benzophenone-2-carboxylic Acid Methyl Ester, Methyl Benzophenone-2-carboxylate, 2-Benzoylbenzoic Acid Methyl Ester |
| PubChem CID | 11816 |
| IUPAC Name | methyl 2-benzoylbenzoate |
| SMILES | COC(=O)C1=CC=CC=C1C(=O)C2=CC=CC=C2 |
Chemical Identifiers
| 606-28-0 | |
| 240.258 | |
| NQSMEZJWJJVYOI-UHFFFAOYSA-N | |
| 11816 | |
| COC(=O)C1=CC=CC=C1C(=O)C2=CC=CC=C2 |
| C15H12O3 | |
| MFCD00017187 | |
| Benzophenone-2-carboxylic Acid Methyl Ester, Methyl Benzophenone-2-carboxylate, 2-Benzoylbenzoic Acid Methyl Ester | |
| methyl 2-benzoylbenzoate |
Specifications
| 606-28-0 | |
| White | |
| C15H12O3 | |
| 25 g | |
| NQSMEZJWJJVYOI-UHFFFAOYSA-N | |
| methyl 2-benzoylbenzoate | |
| 11816 | |
| ≥98.0% (GC) | |
| Methyl 2-Benzoylbenzoate |
| 52°C | |
| 352°C | |
| MFCD00017187 | |
| Benzophenone-2-carboxylic Acid Methyl Ester, Methyl Benzophenone-2-carboxylate, 2-Benzoylbenzoic Acid Methyl Ester | |
| COC(=O)C1=CC=CC=C1C(=O)C2=CC=CC=C2 | |
| 240.258 | |
| 240.26 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0267
TSCA : Yes