missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Benzoylbenzoate 98.0+%, TCI America™

Catalog No. b1231500g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1231500G 500 g
B123125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1231500G Supplier TCI America Supplier No. B1231500G
Only null left

Chemical Identifiers

CAS 606-28-0
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.258
MDL Number MFCD00017187
InChI Key NQSMEZJWJJVYOI-UHFFFAOYSA-N
Synonym Benzophenone-2-carboxylic Acid Methyl Ester, Methyl Benzophenone-2-carboxylate, 2-Benzoylbenzoic Acid Methyl Ester
PubChem CID 11816
IUPAC Name methyl 2-benzoylbenzoate
SMILES COC(=O)C1=CC=CC=C1C(=O)C2=CC=CC=C2

Specifications

Melting Point 52°C
Color White
Boiling Point 352°C
Quantity 500 g
Formula Weight 240.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Benzoylbenzoate
EINECSNumber (4)-0267
TSCA Yes