missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Amino-5-methylbenzoate 98.0+%, TCI America™

Catalog No. M25415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25415G 5 g
M254125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25415G Supplier TCI America Supplier No. M25415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18595-16-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00661547
InChI Key MDHYFUPTSWXVIA-UHFFFAOYSA-N
Synonym 2-Amino-5-methylbenzoic Acid Methyl Ester, Methyl 6-Amino-m-toluate, 6-Amino-m-toluic Acid Methyl Ester, Methyl 5-Methylanthranilate, 5-Methylanthranilic Acid Methyl Ester
PubChem CID 1201104
IUPAC Name methyl 2-amino-5-methylbenzoate
SMILES COC(=O)C1=C(N)C=CC(C)=C1

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Amino-5-methylbenzoate