CAS RN 18595-16-9
IUPAC Name:
methyl 2-amino-5-methylbenzoate
Synonyms:
methyl 2-amino-5-methylbenzoate
Molecular Weight (g/mol):
165.19
Molecular Formula:
C9H11NO2
InChi Key:
MDHYFUPTSWXVIA-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(N)C=CC(C)=C1
1
–
1
of
1
results
1
Methyl 2-Amino-5-methylbenzoate 98.0+%, TCI America™
CAS: 18595-16-9 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.19 MDL Number: MFCD00661547 InChI Key: MDHYFUPTSWXVIA-UHFFFAOYSA-N Synonym: 2-Amino-5-methylbenzoic Acid Methyl Ester, Methyl 6-Amino-m-toluate, 6-Amino-m-toluic Acid Methyl Ester, Methyl 5-Methylanthranilate, 5-Methylanthranilic Acid Methyl Ester PubChem CID: 1201104 IUPAC Name: methyl 2-amino-5-methylbenzoate SMILES: COC(=O)C1=C(N)C=CC(C)=C1