missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3-methylbenzoate, 98%

Catalog No. AAH6144814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6144814 25 g
AAH6144822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6144814 Supplier Thermo Scientific Chemicals Supplier No. H6144814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22223-49-0 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22223-49-0
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD06200918
InChI Key VSFYTPXXMLJNAU-UHFFFAOYSA-N
Synonym methyl 3-methylanthranilate, 2-amino-3-methylbenzoic acid methyl ester, benzoic acid, 2-amino-3-methyl-, methyl ester, methyl 2-amino-3-methylbenzenecarboxylate, 2-amino-3-methyl-benzoic acid methyl ester, methyl2-amino-3-methylbenzoate, pubchem10929, acmc-209ftn, methyl 2-amino-m-toluate, ksc495o6t
PubChem CID 2763406
IUPAC Name methyl 2-amino-3-methylbenzoate
SMILES CC1=CC=CC(=C1N)C(=O)OC

Specifications

Melting Point 25°C to 29°C
Color Pink
Boiling Point 153°C (23 mm)
Quantity 25 g
Formula Weight 165.19
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-3-methylbenzoate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 244-847-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only