missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-Dihydroxybenzoate 98.0+%, TCI America™

Catalog No. D14615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14615G 5 g
D146125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14615G Supplier TCI America Supplier No. D14615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2150-45-0
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00059619
InChI Key WCQZCKUNZVMBDC-UHFFFAOYSA-N
Synonym 2,6-dihydroxybenzoic acid methyl ester, benzoic acid, 2,6-dihydroxy-, methyl ester, .gamma.-resorcylic acid, methyl ester, acmc-209fl9, 2-methoxycarbonyl resorcinol, methyl 2,6-dihydroxy benzoate, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate, 2-methoxycarbonyl benzene-1,3-diol
PubChem CID 519869
IUPAC Name methyl 2,6-dihydroxybenzoate
SMILES COC(=O)C1=C(O)C=CC=C1O

Specifications

Melting Point 70°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,6-Dihydroxybenzoate