CAS RN 2150-45-0
IUPAC Name:
methyl 2,6-dihydroxybenzoate
Synonyms:
methyl 2,6-dihydroxybenzoate
Molecular Weight (g/mol):
168.15
Molecular Formula:
C8H8O4
InChi Key:
WCQZCKUNZVMBDC-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(O)C=CC=C1O
1
–
1
of
1
results
1
Methyl 2,6-Dihydroxybenzoate 98.0+%, TCI America™
CAS: 2150-45-0 Molecular Formula: C8H8O4 Molecular Weight (g/mol): 168.15 MDL Number: MFCD00059619 InChI Key: WCQZCKUNZVMBDC-UHFFFAOYSA-N Synonym: 2,6-dihydroxybenzoic acid methyl ester,benzoic acid, 2,6-dihydroxy-, methyl ester,.gamma.-resorcylic acid, methyl ester,acmc-209fl9,2-methoxycarbonyl resorcinol,methyl 2,6-dihydroxy benzoate,gamma-resorcylic acid methyl ester,methyl 2,6-dihydroxybenzoate,methyl-2,6-dihydroxybenzoate,2-methoxycarbonyl benzene-1,3-diol PubChem CID: 519869 IUPAC Name: methyl 2,6-dihydroxybenzoate SMILES: COC(=O)C1=C(O)C=CC=C1O