missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-Difluoro-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7133403
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M27491G 1 g
M27495G 5 g
2 options

Catalog No. M27491G

Supplier: TCI America M27491G

Only null left

Chemical Identifiers

CAS 84832-01-9
Molecular Formula C8H5F2NO4
Molecular Weight (g/mol) 217.128
MDL Number MFCD07368850
InChI Key CIHHBTMZOLRCRL-UHFFFAOYSA-N
PubChem CID 43449035
IUPAC Name methyl 2,6-difluoro-3-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1F)[N+](=O)[O-])F

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 151°C
Quantity 1 g
Synonym 2,6-difluoro-3-nitrobenzoic acid methyl ester, benzoic acid,2,6-difluoro-3-nitro-, methyl ester, benzoic acid, 2,6-difluoro-3-nitro-, methyl ester, acmc-209pwt, methyl2,6-difluoro-3-nitrobenzoate, 2,6-difluoro-3-nitro-benzoic acid methyl ester
Formula Weight 217.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,6-Difluoro-3-nitrobenzoate