accessibility menu, dialog, popup

UserName

CAS RN 84832-01-9

CH3OOFNOOF

CAS RN 84832-01-9

IUPAC Name: methyl 2,6-difluoro-3-nitrobenzoate
Synonyms: methyl 2,6-difluoro-3-nitrobenzoate
Molecular Weight (g/mol): 217.13
Molecular Formula: C8H5F2NO4
InChi Key: CIHHBTMZOLRCRL-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C=CC(=C1F)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results