missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-Dihydroxybenzoate 98.0+%, TCI America™

Catalog No. p-7132961
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20065G 5 g
M200625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20065G Supplier TCI America Supplier No. M20065G
Only null left

Chemical Identifiers

CAS 2150-46-1
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00016464
InChI Key XGDPKUKRQHHZTH-UHFFFAOYSA-N
Synonym methyl gentisate, gentisic acid methyl ester, methyl2,5-dihydroxybenzoate, benzoic acid, 2,5-dihydroxy-, methyl ester, methyl dihydroxybenzoate, 2,5-dihydroxybenzoic acid methyl ester, gentisic acid, methyl ester, methyl-2,5-dihydroxybenzoate, unii-ls28b004km, rarechem al bf 0043
PubChem CID 75077
IUPAC Name methyl 2,5-dihydroxybenzoate
SMILES COC(=O)C1=CC(O)=CC=C1O

Specifications

Melting Point 87°C
Boiling Point 136°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,5-Dihydroxybenzoate