missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-dihydroxybenzoate, 98%

Catalog No. AAA1040706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1040706 5 g
AAA1040714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1040706 Supplier Thermo Scientific Chemicals Supplier No. A1040706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2150-46-1 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2150-46-1
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00016464
InChI Key XGDPKUKRQHHZTH-UHFFFAOYSA-N
Synonym methyl gentisate, gentisic acid methyl ester, methyl2,5-dihydroxybenzoate, benzoic acid, 2,5-dihydroxy-, methyl ester, methyl dihydroxybenzoate, 2,5-dihydroxybenzoic acid methyl ester, gentisic acid, methyl ester, methyl-2,5-dihydroxybenzoate, unii-ls28b004km, rarechem al bf 0043
PubChem CID 75077
IUPAC Name methyl 2,5-dihydroxybenzoate
SMILES COC(=O)C1=CC(O)=CC=C1O

Specifications

Melting Point 86°C to 87°C
Quantity 5 g
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material Methyl 2,5-dihydroxybenzoate
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 218-427-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only