missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-difluoro-4-nitrobenzoate, 95%

Catalog No. AAH6347206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6347206 5 g
AAH6347203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6347206 Supplier Thermo Scientific Chemicals Supplier No. H6347206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 924868-81-5 | C8H5F2NO4 | 217.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 924868-81-5
Molecular Formula C8H5F2NO4
Molecular Weight (g/mol) 217.13
MDL Number MFCD08689670
InChI Key XBUVRWIIEREYFL-UHFFFAOYSA-N
Synonym methyl 2,5-difluoro-4-nitrobenzenecarboxylate, methyl2,5-difluoro-4-nitrobenzoate, benzoic acid, 2,5-difluoro-4-nitro-, methyl ester, methyldifluoronitrobenzenecarboxylate, methyl-2,5-difluoro-4-nitrobenzoate, 2,5-difluoro-4-methoxycarbonyl nitrobenzene, 2,5-difluoro-4-nitro-benzoic acid methyl ester
PubChem CID 18526058
IUPAC Name methyl 2,5-difluoro-4-nitrobenzoate
SMILES COC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O

Specifications

Quantity 5 g
Formula Weight 217.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 2,5-difluoro-4-nitrobenzoate
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only