accessibility menu, dialog, popup

UserName

CAS RN 924868-81-5

CH3OOFNOOF

CAS RN 924868-81-5

IUPAC Name: methyl 2,5-difluoro-4-nitrobenzoate
Synonyms: methyl 2,5-difluoro-4-nitrobenzoate
Molecular Weight (g/mol): 217.13
Molecular Formula: C8H5F2NO4
InChi Key: XBUVRWIIEREYFL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results