missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-dichlorobenzoate, 99%

Catalog No. p-7026480
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2248909 10 g
AAB2248918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2248909 Supplier Thermo Scientific Chemicals Supplier No. B2248909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2905-69-3 | C8H6Cl2O2 | 205.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-69-3
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00000606
InChI Key SPJQBGGHUDNAIC-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzoic acid methyl ester, benzoic acid, 2,5-dichloro-, methyl ester, methyl-2,5-dichlorobenzoate, unii-j07555o4he, benzoic acid, dichloro-, methyl ester, pubchem3714, acmc-1bhkt, 2,5-dcbme, rarechem al bf 0427, methyl 2,5-bis chloranyl benzoate
PubChem CID 17947
IUPAC Name methyl 2,5-dichlorobenzoate
SMILES COC(=O)C1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 37°C to 40°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2251582
Formula Weight 205.04
Percent Purity 99%
Chemical Name or Material Methyl 2,5-dichlorobenzoate
Hazard Category H302-H336
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 220-815-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only