accessibility menu, dialog, popup

UserName

CAS RN 2905-69-3

IUPAC Name: methyl 2,5-dichlorobenzoate
Synonyms: methyl 2,5-dichlorobenzoate
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: SPJQBGGHUDNAIC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results