missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-Dibromobenzoate 98.0+%, TCI America™

Catalog No. p-7127437
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26735G 5 g
D267325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26735G Supplier TCI America Supplier No. D26735G

Chemical Identifiers

CAS 57381-43-8
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.942
MDL Number MFCD00144757
InChI Key RBCUIRIGTNHLPS-UHFFFAOYSA-N
Synonym 2,5-dibromobenzoic acid methyl ester, benzoic acid, 2,5-dibromo-, methyl ester, pubchem3972, acmc-1aybl, #, methyl 2,5-bis bromanyl benzoate, methyl 2,5-dibromobenzoate, benzoic acid,2,5-dibromo-, methyl ester, 3,5-diamino-2-methyl benzoic acid
PubChem CID 554523
IUPAC Name methyl 2,5-dibromobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Br)Br

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 5 g
Formula Weight 293.94
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,5-Dibromobenzoate