accessibility menu, dialog, popup

UserName

CAS RN 57381-43-8

IUPAC Name: methyl 2,5-dibromobenzoate
Synonyms: methyl 2,5-dibromobenzoate
Molecular Weight (g/mol): 293.94
Molecular Formula: C8H6Br2O2
InChi Key: RBCUIRIGTNHLPS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results