missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetate 98.0+%, TCI America™

Catalog No. H1049100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1049100MG 100 mg
H10491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1049100MG Supplier TCI America Supplier No. H1049100MG
Only null left

Chemical Identifiers

CAS 308815-81-8
Molecular Formula C10H11NO6
Molecular Weight (g/mol) 241.20
MDL Number MFCD04117920
InChI Key HNFVHTJFCGBTSS-UHFFFAOYSA-N
Synonym 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetic Acid Methyl Ester
PubChem CID 43137771
IUPAC Name methyl 2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate
SMILES COC(=O)COC1=C(C=C(CO)C=C1)[N+]([O-])=O

Specifications

Quantity 100 mg
Formula Weight 241.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetate