accessibility menu, dialog, popup

UserName

H3COOOOHNOO

CAS RN 308815-81-8

IUPAC Name: methyl 2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate
Synonyms: methyl 2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate
Molecular Weight (g/mol): 241.2
Molecular Formula: C10H11NO6
InChi Key: HNFVHTJFCGBTSS-UHFFFAOYSA-N
SMILES: COC(=O)COC1=C(C=C(CO)C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results