missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,4-Dihydroxybenzoate 98.0+%, TCI America™
$38.51 - $142.46
Chemical Identifiers
| CAS | 2150-47-2 |
|---|---|
| Molecular Formula | C8H8O4 |
| Molecular Weight (g/mol) | 168.148 |
| MDL Number | MFCD00002276 |
| InChI Key | IIFCLXHRIYTHPV-UHFFFAOYSA-N |
| Synonym | 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate |
| PubChem CID | 16523 |
| IUPAC Name | methyl 2,4-dihydroxybenzoate |
| SMILES | COC(=O)C1=C(C=C(C=C1)O)O |
Chemical Identifiers
| 2150-47-2 | |
| 168.148 | |
| IIFCLXHRIYTHPV-UHFFFAOYSA-N | |
| 16523 | |
| COC(=O)C1=C(C=C(C=C1)O)O |
| C8H8O4 | |
| MFCD00002276 | |
| 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate | |
| methyl 2,4-dihydroxybenzoate |
Specifications
| 2150-47-2 | |
| White | |
| MFCD00002276 | |
| 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate | |
| COC(=O)C1=C(C=C(C=C1)O)O | |
| 168.148 | |
| 168.15 | |
| Crystalline Powder |
| 119°C | |
| C8H8O4 | |
| 5 g | |
| IIFCLXHRIYTHPV-UHFFFAOYSA-N | |
| methyl 2,4-dihydroxybenzoate | |
| 16523 | |
| ≥98.0% (GC,T) | |
| Methyl 2,4-Dihydroxybenzoate |
Safety and Handling
TSCA : Yes