missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,4-Dihydroxybenzoate 98.0+%, TCI America™
Supplier: TCI America M17965G
Specifications
| Methyl 2,4-Dihydroxybenzoate | |
| 119°C | |
| C8H8O4 | |
| 5 g | |
| IIFCLXHRIYTHPV-UHFFFAOYSA-N | |
| methyl 2,4-dihydroxybenzoate | |
| 16523 | |
| ≥98.0% (GC,T) |
| 2150-47-2 | |
| White | |
| MFCD00002276 | |
| 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate | |
| COC(=O)C1=C(C=C(C=C1)O)O | |
| 168.148 | |
| 168.15 | |
| Crystalline Powder |
Chemical Identifiers
| 2150-47-2 | |
| 168.148 | |
| IIFCLXHRIYTHPV-UHFFFAOYSA-N | |
| 16523 | |
| COC(=O)C1=C(C=C(C=C1)O)O |
| C8H8O4 | |
| MFCD00002276 | |
| 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate | |
| methyl 2,4-dihydroxybenzoate |
Safety and Handling
TSCA : Yes