missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-[4-(Bromomethyl)phenyl]benzoate 98.0+%, TCI America™

Catalog No. M18065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18065G 5 g
M180625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18065G Supplier TCI America Supplier No. M18065G

Chemical Identifiers

CAS 114772-38-2
Molecular Formula C15H13BrO2
Molecular Weight (g/mol) 305.171
MDL Number MFCD06200816
InChI Key RMXGTMRDXKUUDJ-UHFFFAOYSA-N
PubChem CID 11012181
IUPAC Name methyl 2-[4-(bromomethyl)phenyl]benzoate
SMILES COC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 5 g
Synonym 2-[4-(Bromomethyl)phenyl]benzoic Acid Methyl Ester, Methyl 4′C-(Bromomethyl)biphenyl-2-carboxylate, 4′C-(Bromomethyl)biphenyl-2-carboxylic Acid Methyl Ester
Formula Weight 305.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-[4-(Bromomethyl)phenyl]benzoate