accessibility menu, dialog, popup

UserName

CH3OOBr

CAS RN 114772-38-2

IUPAC Name: methyl 4'-(bromomethyl)-[1,1'-biphenyl]-2-carboxylate
Synonyms: methyl 4'-(bromomethyl)-[1,1'-biphenyl]-2-carboxylate
Molecular Weight (g/mol): 305.17
Molecular Formula: C15H13BrO2
InChi Key: RMXGTMRDXKUUDJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C1=CC=C(CBr)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results