missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,3:4,6-Di-O-benzylidene-alpha-D-mannopyranoside 98.0+%, TCI America™
Supplier: TCI America M206125G
Specifications
| Methyl 2,3:4,6-Di-O-benzylidene-alpha-D-mannopyranoside | |
| 185°C | |
| C21H22O6 | |
| 25 g | |
| COC1C2C(C3C(O1)COC(O3)C4=CC=CC=C4)OC(O2)C5=CC=CC=C5 | |
| 370.401 | |
| 370.40 | |
| Crystalline Powder |
| 4148-71-4 | |
| White | |
| MFCD00192286 | |
| VGLOFWHLPMVSBF-JNXOPKTKSA-N | |
| (2S,4S,5aR,8R,9aR,9bS)-4-methoxy-2,8-diphenyl-3a,4,5a,6,9a,9b-hexahydro-[1,3]dioxolo[2,3]pyrano[2,4-d][1,3]dioxine | |
| 71750292 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 4148-71-4 | |
| 370.401 | |
| VGLOFWHLPMVSBF-JNXOPKTKSA-N | |
| (2S,4S,5aR,8R,9aR,9bS)-4-methoxy-2,8-diphenyl-3a,4,5a,6,9a,9b-hexahydro-[1,3]dioxolo[2,3]pyrano[2,4-d][1,3]dioxine |
| C21H22O6 | |
| MFCD00192286 | |
| 71750292 | |
| COC1C2C(C3C(O1)COC(O3)C4=CC=CC=C4)OC(O2)C5=CC=CC=C5 |
Safety and Handling
TSCA : No