accessibility menu, dialog, popup

UserName

CAS RN 4148-71-4

IUPAC Name: 7-methoxy-4,12-diphenyl-3,5,8,11,13-pentaoxatricyclo[7.4.0.0²,⁶]tridecane
Synonyms: 7-methoxy-4,12-diphenyl-3,5,8,11,13-pentaoxatricyclo[7.4.0.0²,⁶]tridecane
Molecular Weight (g/mol): 370.4
Molecular Formula: C21H22O6
InChi Key: VGLOFWHLPMVSBF-UHFFFAOYNA-N
SMILES: COC1OC2COC(OC2C2OC(OC12)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results