missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3:4,6-Di-O-benzylidene-alpha-D-mannopyranoside 98.0+%, TCI America™

Catalog No. p-7132996
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20615G 5 g
M206125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20615G Supplier TCI America Supplier No. M20615G
Only null left

Chemical Identifiers

CAS 4148-71-4
Molecular Formula C21H22O6
Molecular Weight (g/mol) 370.401
MDL Number MFCD00192286
InChI Key VGLOFWHLPMVSBF-JNXOPKTKSA-N
PubChem CID 71750292
IUPAC Name (2S,4S,5aR,8R,9aR,9bS)-4-methoxy-2,8-diphenyl-3a,4,5a,6,9a,9b-hexahydro-[1,3]dioxolo[2,3]pyrano[2,4-d][1,3]dioxine
SMILES COC1C2C(C3C(O1)COC(O3)C4=CC=CC=C4)OC(O2)C5=CC=CC=C5

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 370.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3:4,6-Di-O-benzylidene-alpha-D-mannopyranoside