missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 10-Undecenoate 96.0+%, TCI America™
Supplier: TCI America U0036100ML
Specifications
| Methyl 10-Undecenoate | |
| Yellow | |
| C12H22O2 | |
| 100 mL | |
| KISVAASFGZJBCY-UHFFFAOYSA-N | |
| methyl undec-10-enoate | |
| 8138 | |
| 198.31 | |
| Liquid |
| 111-81-9 | |
| 248°C | |
| MFCD00016689 | |
| 10-Undecenoic Acid Methyl Ester | |
| COC(=O)CCCCCCCCC=C | |
| 198.306 | |
| CHEBI:87493 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 111-81-9 | |
| 198.306 | |
| KISVAASFGZJBCY-UHFFFAOYSA-N | |
| 8138 | |
| methyl undec-10-enoate |
| C12H22O2 | |
| MFCD00016689 | |
| 10-Undecenoic Acid Methyl Ester | |
| CHEBI:87493 | |
| COC(=O)CCCCCCCCC=C |
Safety and Handling
TSCA : Yes