missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 10-Undecenoate 96.0+%, TCI America™
$69.31 - $336.90
Chemical Identifiers
| CAS | 111-81-9 |
|---|---|
| Molecular Formula | C12H22O2 |
| Molecular Weight (g/mol) | 198.306 |
| MDL Number | MFCD00016689 |
| InChI Key | KISVAASFGZJBCY-UHFFFAOYSA-N |
| Synonym | 10-Undecenoic Acid Methyl Ester |
| PubChem CID | 8138 |
| ChEBI | CHEBI:87493 |
| IUPAC Name | methyl undec-10-enoate |
| SMILES | COC(=O)CCCCCCCCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
U003625ML
|
TCI America
U003625ML |
25 mL |
Each for $69.31
|
|
|||||
|
U0036100ML
|
TCI America
U0036100ML |
100 mL |
Each for $150.16
|
|
|||||
|
U0036500ML
|
TCI America
U0036500ML |
500 mL |
Each for $336.90
|
|
|||||
Chemical Identifiers
| 111-81-9 | |
| 198.306 | |
| KISVAASFGZJBCY-UHFFFAOYSA-N | |
| 8138 | |
| methyl undec-10-enoate |
| C12H22O2 | |
| MFCD00016689 | |
| 10-Undecenoic Acid Methyl Ester | |
| CHEBI:87493 | |
| COC(=O)CCCCCCCCC=C |
Specifications
| 111-81-9 | |
| 248°C | |
| MFCD00016689 | |
| 10-Undecenoic Acid Methyl Ester | |
| COC(=O)CCCCCCCCC=C | |
| 198.306 | |
| CHEBI:87493 | |
| ≥96.0% (GC) | |
| Methyl 10-Undecenoate |
| Yellow | |
| C12H22O2 | |
| 25 mL | |
| KISVAASFGZJBCY-UHFFFAOYSA-N | |
| methyl undec-10-enoate | |
| 8138 | |
| 198.31 | |
| Liquid |
Safety and Handling
TSCA : Yes