missing translation for 'onlineSavingsMsg'
Learn More

Methoxytrimethylsilane 98.0+%, TCI America™

Supplier:  TCI America M0688250ML

 View more versions of this product

Catalog No. M0688250ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methoxytrimethylsilane
Colorless
C4H12OSi
250 mL
trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn
CO[Si](C)(C)C
104.22
104.22
Liquid
1825-61-2
57°C
MFCD00008344,MFCD03458977
1993
POPACFLNWGUDSR-UHFFFAOYSA-N
methoxytrimethylsilane
15771
≥98.0% (GC)

Chemical Identifiers

1825-61-2
104.22
POPACFLNWGUDSR-UHFFFAOYSA-N
15771
CO[Si](C)(C)C
C4H12OSi
MFCD00008344,MFCD03458977
trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn
methoxytrimethylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2052&(2)-2053

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions