missing translation for 'onlineSavingsMsg'
Learn More

Methoxytrimethylsilane 98.0+%, TCI America™

Catalog No. p-7132301
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M068825ML 25 mL
M0688250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M068825ML Supplier TCI America Supplier No. M068825ML

Chemical Identifiers

CAS 1825-61-2
Molecular Formula C4H12OSi
Molecular Weight (g/mol) 104.22
MDL Number MFCD00008344,MFCD03458977
InChI Key POPACFLNWGUDSR-UHFFFAOYSA-N
Synonym trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn
PubChem CID 15771
IUPAC Name methoxytrimethylsilane
SMILES CO[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 57°C
Quantity 25 mL
UN Number 1993
Formula Weight 104.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxytrimethylsilane
EINECSNumber (2)-2052&(2)-2053
TSCA Yes