missing translation for 'onlineSavingsMsg'
Learn More

Methoxymethyl Phenyl Sulfone 98.0+%, TCI America™

Catalog No. p-7133336
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M26291G 1 g
M26295G 5 g
2 options

Catalog No. M26291G

Supplier: TCI America M26291G

Only null left

Chemical Identifiers

CAS 15251-78-2
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.225
InChI Key MZIVRTQIEKRAPI-UHFFFAOYSA-N
PubChem CID 366072
IUPAC Name methoxymethylsulfonylbenzene
SMILES COCS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 73°C
Color White
Quantity 1 g
Formula Weight 186.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methoxymethyl Phenyl Sulfone