accessibility menu, dialog, popup

UserName

CAS RN 15251-78-2

CH3OSOO

CAS RN 15251-78-2

IUPAC Name: methoxymethanesulfonylbenzene
Synonyms: methoxymethanesulfonylbenzene
Molecular Weight (g/mol): 186.23
Molecular Formula: C8H10O3S
InChi Key: MZIVRTQIEKRAPI-UHFFFAOYSA-N
SMILES: COCS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results