missing translation for 'onlineSavingsMsg'
Learn More

Methoxymethyl Phenyl Sulfide 96.0+%, TCI America™

Catalog No. M12165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12165G Supplier TCI America Supplier No. M12165G
Only null left

Chemical Identifiers

CAS 13865-50-4
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00008496
InChI Key QPXQVNXSQCRWEV-UHFFFAOYSA-N
Synonym alpha-Methoxythioanisole, Methyl Phenylthiomethyl Ether
PubChem CID 542598
IUPAC Name methoxymethylsulfanylbenzene
SMILES COCSC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 154.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxymethyl Phenyl Sulfide