accessibility menu, dialog, popup

UserName

H3COS

CAS RN 13865-50-4

IUPAC Name: [(methoxymethyl)sulfanyl]benzene
Synonyms: [(methoxymethyl)sulfanyl]benzene
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: QPXQVNXSQCRWEV-UHFFFAOYSA-N
SMILES: COCSC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results