missing translation for 'onlineSavingsMsg'
Learn More

Methoxychlor

Catalog No. p-7058245
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6383906 5 g
AAJ6383914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6383906 Supplier Thermo Scientific Chemicals Supplier No. J6383906
Only null left

Molecule with estrogenic activity | CAS: 72-43-5 | C16H15Cl3O2 | 345.644 g/mol

Molecule with estrogenic activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72-43-5
Molecular Formula C16H15Cl3O2
Molecular Weight (g/mol) 345.644
MDL Number MFCD00000803
InChI Key IAKOZHOLGAGEJT-UHFFFAOYSA-N
Synonym methoxychlor, p,p'-methoxychlor, dmdt, dimethoxy-ddt, methoxy-ddt, methoxcide, metox, maralate, marlate, moxie
PubChem CID 4115
ChEBI CHEBI:6842
IUPAC Name 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl

Specifications

Melting Point 87°C
Color White
Quantity 5 g
UN Number UN3077
Beilstein 2057367
Merck Index 14,5990
Formula Weight 345.65
Physical Form Crystalline Powder
Chemical Name or Material Methoxychlor
Hazard Category H302+H312+H332-H351
Hazard Statement GHS H Statement
H351-H302-H312-H332
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 200-779-9
RTECSNumber KJ3675000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only