missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyacetonitrile 98.0+%, TCI America™
$126.76 - $378.40
Chemical Identifiers
| CAS | 1738-36-9 |
|---|---|
| Molecular Formula | C3H5NO |
| Molecular Weight (g/mol) | 71.08 |
| MDL Number | MFCD00001892 |
| InChI Key | QKPVEISEHYYHRH-UHFFFAOYSA-N |
| PubChem CID | 74442 |
| IUPAC Name | 2-methoxyacetonitrile |
| SMILES | COCC#N |
Chemical Identifiers
| 1738-36-9 | |
| 71.08 | |
| QKPVEISEHYYHRH-UHFFFAOYSA-N | |
| 2-methoxyacetonitrile |
| C3H5NO | |
| MFCD00001892 | |
| 74442 | |
| COCC#N |
Specifications
| 1738-36-9 | |
| 119°C | |
| MFCD00001892 | |
| 1993 | |
| QKPVEISEHYYHRH-UHFFFAOYSA-N | |
| 2-methoxyacetonitrile | |
| 74442 | |
| ≥98.0% (GC) | |
| Methoxyacetonitrile |
| Yellow | |
| C3H5NO | |
| 5 mL | |
| methoxyacetonitrile, acetonitrile, methoxy, acetonitrile, 2-methoxy, ncch2och3, 2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile,2-methoxy, intermediates-zcf02294, acmc-209e7f | |
| COCC#N | |
| 71.08 | |
| 71.08 | |
| Liquid |
Safety and Handling
TSCA : Yes