missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyacetonitrile 98.0+%, TCI America™
Supplier: TCI America M010325ML
Specifications
| Methoxyacetonitrile | |
| Yellow | |
| C3H5NO | |
| 25 mL | |
| methoxyacetonitrile, acetonitrile, methoxy, acetonitrile, 2-methoxy, ncch2och3, 2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile,2-methoxy, intermediates-zcf02294, acmc-209e7f | |
| COCC#N | |
| 71.08 | |
| 71.08 | |
| Liquid |
| 1738-36-9 | |
| 119°C | |
| MFCD00001892 | |
| 1993 | |
| QKPVEISEHYYHRH-UHFFFAOYSA-N | |
| 2-methoxyacetonitrile | |
| 74442 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1738-36-9 | |
| 71.08 | |
| QKPVEISEHYYHRH-UHFFFAOYSA-N | |
| 2-methoxyacetonitrile |
| C3H5NO | |
| MFCD00001892 | |
| 74442 | |
| COCC#N |
Safety and Handling
TSCA : Yes