missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Methoxy-X04

Catalog No. 492050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
492050 50 mg
492010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 492050 missing translation for 'mfr' Tocris Bioscience™ missing translation for 'supplierNo' 4920/50
Only null left

Fluorescent amyloid β detector; brain penetrant

Methoxy-X04 is a fluorescent amyloid β (Aβ) probe for the detection and quantification of plaques, tangles and cerebrovascular amyloid. Methoxy-X04 Displays high in vitro binding affinity (Ki = 26.8 nM); binds selectively to fibrillar β-sheet deposits. Methoxy-X04 is a derivative of Congo Red (Cat. No. 5167). Methoxy-X04 can be used for in vivo Aβ plaque labeling. Methoxy-X04 co-localizes with CD68+ phagosomes in plaque-associated Iba1+ microglia; and it labels molecularly distinct plaque-associated microglia populations. Brain penetrant.

Chemical Identifiers

CAS 863918-78-9
Molecular Formula C23H20O3

Specifications

Quantity 50 mg
Formula Weight 344.4
Purity >98%
Molecular Weight (g/mol) 344.41
Chemical Name or Material Methoxy-X04
Recommended Storage Store at +4°C